missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ISLR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5149777
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5149777 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5149777 Supplier Invitrogen™ Supplier No. PA5149777
Only null left

Rabbit Polyclonal Antibody

ISLR Polyclonal Antibody for Western Blot, IHC (P), ICC/IF

ISLR is a protein coding gene.

Specifications

Antigen ISLR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ISLR
Gene Accession No. O14498
Gene Alias HsT17563; immunoglobulin superfamily containing leucine rich repeat; immunoglobulin superfamily containing leucine-rich repeat protein; ISLR; Meflin; mesenchymal stromal cell- and fibroblast-expressing Linx paralogue; UNQ189/PRO215
Gene Symbols ISLR
Host Species Rabbit
Immunogen Recombinant Immunoglobulin Superfamily Containing Leucine Rich Repeat Protein (ISLR) (Cys19-Phe428).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3671, 686539
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less