missing translation for 'onlineSavingsMsg'
Learn More

ISG15 Monoclonal Antibody (3C2), eBioscience™, Invitrogen™

Catalog No. 5013219
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5013219 25 μg
5013220 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5013219 Supplier Thermo Fisher Scientific Supplier No. 14585780

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Armenian Hamster Monoclonal Antibody

Description: This 3C2 monoclonal antibody reacts with mouse interferon stimulated gene-15 (ISG-15), a 15kDa ubiquitin-like protein (UBL) and cytokine. ISG-15 is translated as a precursor that is subsequently activated by an E1 enzyme, conjugated by an E2 enzyme via the C-terminal LRLRGG motif and ligated by an E3 ligase. The activity of ISG-15 is dependent on its modification of target proteins at specific lysine residues. Several viruses utilize ISG-15 and ubiquitin deconjugating proteases, while influenza B protein NS1 has been shown to specifically bind ISG-15 thereby blocking its conjugation. In addition to its role as a protein modification, ISG-15 is secreted by monocytes and lymphocytes in response to IFNalpha, beta, or gamma, as well as influenza B virus. In this role as cytokine, it has been shown to induce secretion of IFNgamma and support NK cell proliferation and cytolytic activity. Applications Reported: This 3C2 antibody has been reported for use in immunoblotting (WB). Applications Tested: This 3C2 antibody has been tested by immunoblot of Raw 264.7 cell lysates at 2 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Ubiquitin-like protein ISG15 plays a key role in the innate immune response to viral infection either via its conjugation to a target protein (ISGylation) or via its action as a free or unconjugated protein. It is secreted from monocytes in response to type I interferons and causes natural killer (NK)-cell proliferation and an augmentation of non-MCH (major histocompatibility complex)-restricted cytotoxicity. Synthesis is stimulated by IFN-alpha or IFN-beta or IFN-omega, but not IFN-gamma. ISG15 expression is also induced by overexpression of interferon regulatory factors that participate in transcriptional regulation of IFN genes, and by influenza B virus. ISG15 is secreted by cell lines of monocyte, T-lymphocyte, B-lymphocyte, human fibroblasts, and epithelial origins. Enhancement of NK cell proliferation, augmentation of non-major histocompatibility complex-restricted cytotoxicity, and induction of IFN-gamma from T cells identify ISG15 as a member of the cytokine cascade and suggest that it may be responsible for amplifying and directing some of the immunomodulatory effects of IFN-alpha or IFN-beta. ISG15 has has also been shown to function intracellularly as a ubiquitin homolog. Mutations affecting the gene can result in Immunodeficiency 38, with basal ganglia calcification (IMD38).

Specifications

Antigen ISG15
Applications Western Blot
Classification Monoclonal
Clone 3C2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ISG15
Gene Accession No. Q64339
Gene Alias G1P2; hUCRP; IFI15; IGI15; IMD38; interferon, alpha-inducible protein; interferon, alpha-inducible protein (clone IFI-15K); interferon-induced 15 kDa protein; interferon-induced 15-KDa protein; interferon-induced 17 kDa protein; interferon-induced 17-kDa/15-kDa protein; interferon-stimulated protein (15 kDa); interferon-stimulated protein 15; interferon-stimulated protein, 15 kDa; IP17; Irfp; Isg15; ISG15 ubiquitin like modifier; ISG15 ubiquitin-like modifier; ubiquitin cross-reactive protein; Ubiquitin-like protein ISG15; UCRP
Gene Symbols ISG15
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100038882
Target Species Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only.