missing translation for 'onlineSavingsMsg'
Learn More

Isatoic anhydride, 98%

Catalog No. p-150293
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122481000 100 g Plastic bottle
AC122485000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122481000 Supplier Thermo Scientific Chemicals Supplier No. 122481000
Only null left

CAS: 118-48-9 | C8H5NO3 | 163.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-48-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD00006700
InChI Key TXJUTRJFNRYTHH-UHFFFAOYSA-N
Synonym isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride
PubChem CID 8359
SMILES O=C1NC2=CC=CC=C2C(=O)O1

Specifications

Melting Point 243°C
Color Beige to Brown
Flash Point 308°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 27, 264
Solubility Information Solubility in water: decomposes
IUPAC Name 2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Formula Weight 163.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Isatoic anhydride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protect
EINECSNumber 204-255-0
RTECSNumber DM3100000
TSCA TSCA

RUO – Research Use Only