Learn More
Isatoic anhydride, 97%
CAS: 118-48-9 | C8H5NO3 | 163.13 g/mol
Supplier: Thermo Scientific Chemicals A146030E
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isatoic anhydride | |
| ∼237°C (decomposition) | |
| 308°C (586°F) | |
| MFCD00006700 | |
| 136786 | |
| isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride | |
| O=C1NC2=CC=CC=C2C(=O)O1 | |
| 163.13 | |
| 163.13 |
| 118-48-9 | |
| 1.52 | |
| C8H5NO3 | |
| 2500 g | |
| Moisture sensitive | |
| TXJUTRJFNRYTHH-UHFFFAOYSA-N | |
| 1H-3,1-benzoxazine-2,4-dione | |
| 8359 | |
| 97% |
Chemical Identifiers
| 118-48-9 | |
| 163.13 | |
| TXJUTRJFNRYTHH-UHFFFAOYSA-N | |
| 8359 |
| C8H5NO3 | |
| MFCD00006700 | |
| isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride | |
| O=C1NC2=CC=CC=C2C(=O)O1 |
Safety and Handling
GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
H317-H319
EINECSNumber : 204-255-0
RTECSNumber : DM3100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only