missing translation for 'onlineSavingsMsg'
Learn More

Isatoic anhydride, 97%

Catalog No. AAA1460322
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1460322 100 g
AAA1460336 500 g
AAA146030E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1460322 Supplier Thermo Scientific Chemicals Supplier No. A1460322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 118-48-9 | C8H5NO3 | 163.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-48-9
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD00006700
InChI Key TXJUTRJFNRYTHH-UHFFFAOYSA-N
Synonym isatoic anhydride, 2h-3,1-benzoxazine-2,4 1h-dione, 1h-benzo d 1,3 oxazine-2,4-dione, isatoic acid anhydride, 4h-3,1-benzoxazine-2,4 1h-dione, isato acid anhydride, n-carboxyanthranilic anhydride, unii-r8tfa74y4u, 3,1-benzoxazine-2,4 1h-dione, anthranilic acid, n-carboxy-, cyclic anhydride
PubChem CID 8359
SMILES O=C1NC2=CC=CC=C2C(=O)O1

Specifications

Melting Point ∼237°C (decomposition)
Density 1.52
Flash Point 308°C (586°F)
Quantity 100 g
Beilstein 136786
Sensitivity Moisture sensitive
IUPAC Name 2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Formula Weight 163.13
Percent Purity 97%
Chemical Name or Material Isatoic anhydride
Hazard Category H317-H319
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
EINECSNumber 204-255-0
RTECSNumber DM3100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only