missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Irsogladine Maleate 98.0+%, TCI America™
$141.67 - $715.79
Chemical Identifiers
| CAS | 84504-69-8 |
|---|---|
| Molecular Formula | C13H11Cl2N5O4 |
| Molecular Weight (g/mol) | 372.16 |
| MDL Number | MFCD00873566 |
| InChI Key | PJLVTVAIERNDEQ-BTJKTKAUSA-N |
| Synonym | irsogladine maleate, gaslon, gaslon n, unii-66gvu60epq, 66gvu60epq, 2,4-diamino-6-2,5-dichlorophenyl-s-triazine maleate, 6-2,5-dichlorophenyl-1,3,5-triazine-2,4-diamine maleate, 1,3,5-triazine-2,4-diamine, 6-2,5-dichlorophenyl-, z-2-butenedioate 1:1, dsstox_cid_20234 |
| PubChem CID | 5282435 |
| IUPAC Name | (2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine |
| SMILES | OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl |
Chemical Identifiers
| 84504-69-8 | |
| 372.16 | |
| PJLVTVAIERNDEQ-BTJKTKAUSA-N | |
| 5282435 | |
| OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl |
| C13H11Cl2N5O4 | |
| MFCD00873566 | |
| irsogladine maleate, gaslon, gaslon n, unii-66gvu60epq, 66gvu60epq, 2,4-diamino-6-2,5-dichlorophenyl-s-triazine maleate, 6-2,5-dichlorophenyl-1,3,5-triazine-2,4-diamine maleate, 1,3,5-triazine-2,4-diamine, 6-2,5-dichlorophenyl-, z-2-butenedioate 1:1, dsstox_cid_20234 | |
| (2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine |
Specifications
| 84504-69-8 | |
| C13H11Cl2N5O4 | |
| 1 g | |
| PJLVTVAIERNDEQ-BTJKTKAUSA-N | |
| (2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine | |
| 5282435 | |
| ≥98.0% (HPLC,T) | |
| Irsogladine Maleate |
| White | |
| MFCD00873566 | |
| irsogladine maleate, gaslon, gaslon n, unii-66gvu60epq, 66gvu60epq, 2,4-diamino-6-2,5-dichlorophenyl-s-triazine maleate, 6-2,5-dichlorophenyl-1,3,5-triazine-2,4-diamine maleate, 1,3,5-triazine-2,4-diamine, 6-2,5-dichlorophenyl-, z-2-butenedioate 1:1, dsstox_cid_20234 | |
| OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl | |
| 372.16 | |
| 372.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XY5930000
TSCA : No