missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Hazard Bags
GREENER_CHOICE

Low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.

$360.41 - $469.57

Specifications

Description Five folded liners per double bag
Disposable Single-use
Quantity 50/Cs.
Autoclavable Not Autoclavable
Thickness 0.050 mm
View More Specs

Products 2
Catalog Number Mfr. No. Color Capacity (Metric) Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Color Capacity (Metric) Sterility Price Quantity & Availability  
19263623
GREENER_CHOICE
View Documents Promotion Details
Micronova™
BL224YEIRBB
Yellow 61 L Sterile
Case of 50 Each for $360.41
Only null left
 
19116737
GREENER_CHOICE
View Documents Promotion Details
Micronova™
BL23440REBB
Red 125 L Non-sterile
Case of 50 Each for $469.57
Only null left
 
Description

Description

Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste

  • Low density polyethylene Hazard bags
  • Red or Yellow Biohazard and Hazardous Material bags
  • Biohazard bags are clearly marked with a Biohazard symbol in black ink
  • Hazardous material bags are labeled for waste segregation in the US
  • The 32YE liners work with the BLSSH-1515 BinLiner Stand
  • The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas
Specifications

Specifications

Five folded liners per double bag
50/Cs.
0.050 mm
Biohazard Waste Bag
Single-use
Not Autoclavable
LDPE
15°C to 30°C
Videos
SDS
Documents

Documents