missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Hazard Bags
GREENER_CHOICE

Catalog No. 19263623
Change view
Click to view available options
Color:
Red
Yellow
Capacity (Metric):
61 L
125 L
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Capacity (Metric) Sterility
19263623 Yellow 61 L Sterile
19116737 Red 125 L Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19263623 Supplier Micronova™ Supplier No. BL224YEIRBB

Low density, LDPE hazard bags are designed for containing biomedical and hazardous waste in aseptic and controlled environments.

Micronova Hazard Bags are constructed from FDA compliant, low density, red or yellow polyethylene and can be used with the BLSSH series Micronova BinLiner Stands. The Biohazard bags are designed for containment and disposal of biomedical waste and the Hazardous Material bags are designed for segregation and disposal of hazardous waste

  • Low density polyethylene Hazard bags
  • Red or Yellow Biohazard and Hazardous Material bags
  • Biohazard bags are clearly marked with a Biohazard symbol in black ink
  • Hazardous material bags are labeled for waste segregation in the US
  • The 32YE liners work with the BLSSH-1515 BinLiner Stand
  • The 24YE and 25RE work with the BLSSH-168 BinLiner Stand
  • Smaller bags with a star bottom are ideal for round containers
  • Larger bags are flat sealed
  • Good compatibility with common cleaners
  • Gamma irradiated liners are sterile for aseptic areas

Specifications

Capacity (Metric) 61 L
Certifications/Compliance Certificate of Conformance and Certificate of Irradiation Processing; ISO 9001 Certified Facility Meets ASTM D170910-6 Sterility Assurance Level In-Compliance with AAMI Guidelines for Sterilization.
Description Five folded liners per double bag
Diameter (Metric) Outer 39 cm
Diameter (English) Outer 15.3 in.
Capacity (English) 16 gal.
Color Yellow
Dimensions (L x W) 24 x 36 in.
Disposable Single-use
Height (Metric) 91 cm
Length (Metric) 61 cm
Quantity 50/Cs.
Shape STAR: Durable pleated bottom to fit round containers.
Sterility Sterile
Autoclavable Not Autoclavable
For Use With (Application) Transporting Biomedical Waste (Non-Sharps)
For Use With (Equipment) BLSSH series BinStand or similar container
Length (English) 24 in.
Thickness 0.050 mm
Height (English) 36 in.
Material LDPE
Product Type Biohazard Waste Bag
Temperature (Metric) 15°C to 30°C
Show More Show Less