missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Stearate, TCI America™

Catalog No. S025625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S025625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S025625G Supplier TCI America Supplier No. S025625G
Only null left

Chemical Identifiers

CAS 555-36-2
Molecular Formula C54H105FeO6
Molecular Weight (g/mol) 906.273
MDL Number MFCD00050716
InChI Key XHQSLVIGPHXVAK-UHFFFAOYSA-K
Synonym Ferric Stearate, Stearic Acid Iron(III) Salt
PubChem CID 68388
IUPAC Name iron(3+);octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Fe+3]

Specifications

Quantity 25 g
Formula Weight 906.27
Physical Form Crystalline Powder
Chemical Name or Material Iron(III) Stearate
EINECSNumber (2)-0625
TSCA Yes