accessibility menu, dialog, popup

UserName

CAS RN 555-36-2

FeH3COOH3COOH3COO

CAS RN 555-36-2

IUPAC Name: iron(3+) trioctadecanoate
Synonyms: iron(3+) tris(n-octadecanoate)
Molecular Weight (g/mol): 906.27
Molecular Formula: C54H105FeO6
InChi Key: XHQSLVIGPHXVAK-UHFFFAOYSA-K
SMILES: [Fe+3].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results