missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Oxide, purified, Honeywell Fluka™

Catalog No. p-7071780
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001928 250 g
6001927 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001928 Supplier Honeywell Chemicals Supplier No. 12342250G
Only null left

Purified,≥95%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Quantity 250 g
UN Number NONH for all modes of transport
Formula Weight 159.69g/mol
Percent Purity ≥95%
Grade Purified
Chemical Name or Material Iron(Iii) Oxide
EINECSNumber 215-168-2
RTECSNumber NO7400000