missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) oxide, cosmetic, NanoArc™, Thermo Scientific Chemicals

Nanoarc Cosmetic Iron Oxide Is An Auburn Material offering Unique Cosmetic Qualitites. The Small Particle Size Results in a Strong Color Development Combined With High Translucency, For a Unique Glowing Look

$73.42 - $820.86

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4489514
View Documents
Thermo Scientific Chemicals
04489514
25 g
Each for $73.42
Only null left
 
AA4489522
View Documents
Thermo Scientific Chemicals
04489522
100 g
Each for $206.53
Only null left
 
AA4489536
View Documents
Thermo Scientific Chemicals
04489536
500 g
Each for $820.86
Only null left
 
Description

Description

NanoArc cosmetic iron oxide is an auburn material offering unique cosmetic qualities. The small particle size results in a strong color development combined with high translucency for a unique glowing look.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 30-60m2/g
Specifications

Chemical Identifiers

1309-37-1
159.69
JEIPFZHSYJVQDO-UHFFFAOYSA-N
[Fe][Fe](=O)(=O)=O
Fe2O3
MFCD00011008
trioxodiiron

Specifications

1565°C
1309-37-1
25 g
MFCD00011008
JEIPFZHSYJVQDO-UHFFFAOYSA-N
trioxodiiron
159.69
Red-brown powder
Iron(III) oxide, cosmetic, Sold in collaboration with Nanophase Technologies Corporation
Red to Brown
Powder
Fe2O3
Insoluble in water
[Fe][Fe](=O)(=O)=O
159.69
Odorless
5.24
Videos
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 215-168-2

RTECSNumber : NO7400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only