Learn More
Iron(III) oxide, alpha-phase, nanopowder, 98% (metals basis), Thermo Scientific Chemicals
$174.93 - $1814.74
Chemical Identifiers
| CAS | 1309-37-1 |
|---|---|
| Molecular Formula | Fe2O3 |
| Molecular Weight (g/mol) | 159.69 |
| MDL Number | MFCD00011008 |
| InChI Key | JEIPFZHSYJVQDO-UHFFFAOYSA-N |
| IUPAC Name | trioxodiiron |
| SMILES | [Fe][Fe](=O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4704414
|
Thermo Scientific Chemicals
04704414 |
25 g |
Each for $174.93
|
|
|||||
|
AA4704422
|
Thermo Scientific Chemicals
04704422 |
100 g |
Each for $550.78
|
|
|||||
|
AA4704436
|
Thermo Scientific Chemicals
04704436 |
500 g |
Each for $1,814.74
|
|
|||||
Description
Ferric oxide is used for polishing precious metals and diamonds, as well as in cosmetics. Synthetic iron oxide pigments are one of the most popular color pigments worldwide for coloring concrete. Other uses includes: coating, printing ink, paint, also as coloring agent for building material, rubber, paper-making. In producing ferrites . Also used in recording tapes and pyrotechnics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1309-37-1 | |
| 159.69 | |
| JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
| [Fe][Fe](=O)(=O)=O |
| Fe2O3 | |
| MFCD00011008 | |
| trioxodiiron |
Specifications
| 1565°C | |
| 1309-37-1 | |
| 25 g | |
| MFCD00011008 | |
| It is soluble In Warm Hydrochloric Acid,Slightly Soluble in Sulfuric Acid. | |
| [Fe][Fe](=O)(=O)=O | |
| 159.69 | |
| 98% | |
| 5.24 g/mL | |
| (metals basis) |
| Red to Brown | |
| Powder | |
| Fe2O3 | |
| 14,4028 | |
| JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
| trioxodiiron | |
| 159.69 | |
| Odorless | |
| Iron (III) oxide, alpha-phase |
Safety and Handling
EINECSNumber : 215-168-2
RTECSNumber : NO7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only