Learn More
Iron(III) oxide, 98% (metals basis), Thermo Scientific Chemicals
$117.75 - $1009.75
Chemical Identifiers
| CAS | 1309-37-1 |
|---|---|
| Molecular Formula | Fe2O3 |
| Molecular Weight (g/mol) | 159.69 |
| MDL Number | MFCD00011008 |
| InChI Key | JEIPFZHSYJVQDO-UHFFFAOYSA-N |
| IUPAC Name | trioxodiiron |
| SMILES | [Fe][Fe](=O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA12375A1
|
Thermo Scientific Chemicals
012375A1 |
1 kg |
Each for $117.75
|
|
|||||
|
AA12375A7
|
Thermo Scientific Chemicals
012375A7 |
5 kg |
Each for $291.27
|
|
|||||
|
AA12375C4
|
Thermo Scientific Chemicals
012375C4 |
25 kg |
Each for $1,009.75
|
|
|||||
Description
It has many applications, mainly used in iron industry, pigment, as photocatalyst, medicine and polishing jewels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1309-37-1 | |
| 159.69 | |
| JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
| [Fe][Fe](=O)(=O)=O |
| Fe2O3 | |
| MFCD00011008 | |
| trioxodiiron |
Specifications
| 1565°C | |
| 1309-37-1 | |
| 1 kg | |
| Fe2O3 | |
| Insoluble in water,Soluble in diluted acids,sugar solution and slightly soluble in aq. tartaric acid,citric acid and acetic acid. | |
| [Fe][Fe](=O)(=O)=O | |
| 159.69 | |
| 5.24 |
| Red to Brown | |
| Powder | |
| 98% | |
| MFCD00011008 | |
| JEIPFZHSYJVQDO-UHFFFAOYSA-N | |
| trioxodiiron | |
| 159.69 | |
| Iron (III) Oxide |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber : 215-168-2
RTECSNumber : NO7400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only