missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) nitrate nonahydrate, ACS, 98.0-101.0%

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol

Supplier:  Thermo Scientific Chemicals 033315A3

 View more versions of this product

Catalog No. AA33315A3


Only null left
Explore available promotions

Description

Description

It is used as catalyst for the synthesis of sodium amide from sodium and ammonia, and for etching of silver and silver alloys. Ferric nitrate is used in the synthesis of organically templated iron phosphates, and acts as an efficient catalyst in enamination of beta dicarbonyl compounds to beta enaminones. Ferric nitrate impregnated clays are useful as oxidizing agents. Clayfen, produced from ferric nitrate and montmorillonite, is used for the oxidation of thiols to disulfide, and alcohols to aldehydes. Ferric nitrate is useful for regio- and stereo-selective nitration of various aromatic, aliphatic, and heteroaromatic olefins. This reaction provides nitroolefins with excellent E-selectivity. With Keggin-type heteropoly acids, it selectively oxidizes various hydroxy groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

125°C (decomposition)
47°C
7782-61-8
Crystalline
98.0 to 101.0%
MFCD00149708
Freely soluble in water,alcohol,acetone. Slightly soluble in cold and concentrated HNO3Soluble in water,alcohol and acetone.
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
403.99
403.99 (241.86 Anhydrous)
Acidic
Pale purple
Iron(III) nitrate nonahydrate
For ACS analysis
Purple
2 kg
FeH18N3O18
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
SZQUEWJRBJDHSM-UHFFFAOYSA-N
iron(3+) nonahydrate trinitrate
16211566
ACS
Moisture sensitive
1.684 g/cm3

Chemical Identifiers

7782-61-8
403.99
SZQUEWJRBJDHSM-UHFFFAOYSA-N
16211566
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
FeH18N3O18
MFCD00149708
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
iron(3+) nonahydrate trinitrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : DOT Class: 5.1, Packing Group: III

EINECSNumber : 233-899-5

RTECSNumber : NO7175000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only