Learn More
Iron(III) nitrate nonahydrate, 98+%, ACS reagent
CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol
Supplier: Thermo Scientific Chemicals 423710030
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Iron(III) nitrate nonahydrate | |
| 7782-61-8 | |
| Adhering Crystalline Powder | |
| 98.0 to 101% (Complexometry) | |
| Fe(NO3)3·9H2O | |
| 1466 | |
| ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
| SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
| iron(3+) nonahydrate trinitrate | |
| 16211566 | |
| ≥98% | |
| Plastic Bottle | |
| Negligible | |
| 1.6800g/mL |
| 47.0°C | |
| Gray to Purple | |
| 3 kg | |
| FeH18N3O18 | |
| MFCD00149708 | |
| 15, 4054 | |
| Solubility in water: soluble. Other solubilities: very soluble in ethanol, acetone | |
| O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 403.99 | |
| 404 | |
| ACS Reagent | |
| 1.68 | |
| Decomposition Information: < 100°C |
Chemical Identifiers
| 7782-61-8 | |
| 403.99 | |
| SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
| 16211566 | |
| O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| FeH18N3O18 | |
| MFCD00149708 | |
| ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
| iron(3+) nonahydrate trinitrate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cauti
GHS Signal Word: Warning
RUO – Research Use Only