missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) nitrate nonahydrate, 98+%, ACS reagent

Catalog No. p-4228999
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423715000 500 g Plastic bottle
AC423710030 3 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423715000 Supplier Thermo Scientific Chemicals Supplier No. 423715000
Only null left

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 47.0°C
Color Gray to Purple
Physical Form Adhering Crystalline Powder
Quantity 500 g
Assay Percent Range 98+%
Linear Formula Fe(NO3)3·9H2O
UN Number 1466
Merck Index 15, 4054
Formula Weight 404
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 1.68
Vapor Pressure Negligible
Decomposition Information Decomposition Information: < 100°C
Density 1.6800g/mL
Chemical Name or Material Iron(III) nitrate nonahydrate
Show More Show Less
Hazard Category SIGNAL WORD: DANGER!
Hazard Statement GHS P Statement
Causes serious eye irritation.
Causes skin irritation.
May intensify fire; oxidizer.
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Irritating to eyes, respiratory system and skin. Hygroscopic.
NFPA
Health: 2
Flammability: 1
Instability: 2

RUO – Research Use Only