missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) isopropoxide, 2.5% w/v in isopropanol

Catalog No. AA43024AA
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43024AA 5 mL
AA43024AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43024AA Supplier Thermo Scientific Chemicals Supplier No. 043024AA

CAS: 14995-22-3 | C9H21FeO3 | 233.109 g/mol

Iron(III) isopropoxide is used in nano-materials & thin films, engineering & environmental, catalysis & synthesis and energy conversion & storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 14995-22-3
Molecular Formula C9H21FeO3
Molecular Weight (g/mol) 233.109
MDL Number MFCD00070436
InChI Key QUHDSMAREWXWFM-UHFFFAOYSA-N
Synonym iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe
PubChem CID 11961763
IUPAC Name iron(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3]

Specifications

Color Red to Brown
Density 0.79 g/mL
Boiling Point 82°C
Flash Point 12°C (54°F)
Odor Alcohol-like
Quantity 5 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 233.11
Concentration or Composition (by Analyte or Components) 2.5% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Iron (III) isopropoxide
Show More Show Less
Hazard Category H225-H319-H335-H336
Hazard Statement GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only