Learn More
Iron(III) hexacyanoferrate(II)
Description
Iron(III) hexacyanoferrate(II) is uses extensively as an Iron Oxide dye. Prussian blue is used as a paint and wallpaper printing, chemical coatings, carbon paper and in the plastics industry. as an antidote for poisoning with radioactive cesium or thallium. In the metalworking and mechanical engineering Prussian blue is thinly applied as a paste on metal surfaces in order to assess the quality scraped surfaces can.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Blue |
| Physical Form | Powder |
| Quantity | 25 g |
| Solubility Information | It is practically insoluble in water. |
| Formula Weight | 859.25 |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Iron(III) hexacyanoferrate(II) |
Safety and Handling
| Hazard Category | H301-H311-H332: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. |
| EINECSNumber | 237-875-5 |
| RTECSNumber | LJ8200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only