missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Hexacyanoferrate(II), TCI America™

Catalog No. I084925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I084925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I084925G Supplier TCI America Supplier No. I084925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14038-43-8
Molecular Formula C18Fe7N18
Molecular Weight (g/mol) 859.239
MDL Number MFCD00135663
InChI Key DCYOBGZUOMKFPA-UHFFFAOYSA-N
Synonym prussian blue, ferric ferrocyanide, iron iii ferrocyanide, iron iii hexacyanoferrate ii, ferric hexacyanoferrate, parisian blue, berlin blue, ferrocin, preussischblau, turnbulls blau
PubChem CID 2724251
IUPAC Name iron(2+);iron(3+);octadecacyanide
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[Fe+2].[Fe+2].[Fe+2].[Fe+3].[Fe+3].[Fe+3].[Fe+3]

Specifications

Color Blue-Purple
Quantity 25 g
Formula Weight 859.24
Physical Form Crystalline Powder
Chemical Name or Material Iron(III) Hexacyanoferrate(II)
RTECSNumber LJ8200000
TSCA Yes