Learn More
Iron(III) ethoxide, 99.6% (metals basis)
Precursor for Sol-Gel processes | CAS: 5058-42-4 | C6H15FeO3 | 191.028 g/mol
Supplier: Thermo Scientific Chemicals 04186304
Description
Precursor for Sol-Gel processes.Iron(III) ethoxide is used as a precursor for inorganic compound iron oxide. It is used in the preparation of iron-platinum nanoparticles by pyrolysis with platinum(II) acetylacetonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
| Iron(III) ethoxide | |
| 120°C | |
| 155°C (0.1 mmHg) | |
| (metals basis) | |
| Fe(OCH2CH3)3 | |
| 2 g | |
| Moisture sensitive | |
| Soluble in anhydrous hexane,alcohols;Miscible with hexane,benzene and ethanol. | |
| CC[O-].CC[O-].CC[O-].[Fe+3] | |
| 191.028 | |
| 191.03 | |
| Chunks and/or Powder |
| 5058-42-4 | |
| Brown to Gray | |
| Odorless | |
| C6H15FeO3 | |
| MFCD00078028 | |
| UN3181 | |
| iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg | |
| JTPUGUWXHGEEHW-UHFFFAOYSA-N | |
| ethanolate;iron(3+) | |
| 6452222 | |
| 99.6% |
Chemical Identifiers
| 5058-42-4 | |
| 191.028 | |
| JTPUGUWXHGEEHW-UHFFFAOYSA-N | |
| 6452222 | |
| CC[O-].CC[O-].CC[O-].[Fe+3] |
| C6H15FeO3 | |
| MFCD00078028 | |
| iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg | |
| ethanolate;iron(3+) |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber : 225-760-2
TSCA : No
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only