missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) ethoxide, 99.6% (metals basis)

Catalog No. AA4186302
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4186302 0.5 g
AA4186304 2 g
AA4186309 10 g
AA4186318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4186302 Supplier Thermo Scientific Chemicals Supplier No. 04186302

Precursor for Sol-Gel processes | CAS: 5058-42-4 | C6H15FeO3 | 191.028 g/mol

Precursor for Sol-Gel processes.Iron(III) ethoxide is used as a precursor for inorganic compound iron oxide. It is used in the preparation of iron-platinum nanoparticles by pyrolysis with platinum(II) acetylacetonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon

Chemical Identifiers

CAS 5058-42-4
Molecular Formula C6H15FeO3
Molecular Weight (g/mol) 191.028
MDL Number MFCD00078028
InChI Key JTPUGUWXHGEEHW-UHFFFAOYSA-N
Synonym iron iii ethoxide, iron 3+ ethanolate, triethoxyiron iii, ethanolate; iron 3+, acmc-1alnr, iron 3+ tris ethoxide, iron 3+ ion tris ethoxide, iron iii ethoxide trace metals basis 500mg
PubChem CID 6452222
IUPAC Name ethanolate;iron(3+)
SMILES CC[O-].CC[O-].CC[O-].[Fe+3]

Specifications

Melting Point 120°C
Color Brown to Gray
Boiling Point 155°C (0.1 mmHg)
Odor Odorless
Linear Formula Fe(OCH2CH3)3
Quantity 0.5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Soluble in anhydrous hexane,alcohols;Miscible with hexane,benzene and ethanol.
Formula Weight 191.03
Percent Purity 99.6%
Physical Form Chunks and/or Powder
Assay (metals basis)
Chemical Name or Material Iron(III) ethoxide
Show More Show Less
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber 225-760-2
TSCA No
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only