missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Dimethyldithiocarbamate 97.0+%, TCI America™
$91.33 - $372.81
Chemical Identifiers
| CAS | 14484-64-1 |
|---|---|
| Molecular Formula | C9H18FeN3S6 |
| Molecular Weight (g/mol) | 416.47 |
| MDL Number | MFCD00067269 |
| InChI Key | WHDGWKAJBYRJJL-UHFFFAOYSA-K |
| Synonym | fuklasin, trimanzone, cormate, fermacide, fermate, ferradow, hexaferb, hokmate, trifungol, ferbam |
| PubChem CID | 86289068 |
| IUPAC Name | iron(3+) tris((dimethylcarbamothioyl)sulfanide) |
| SMILES | [Fe+3].CN(C)C([S-])=S.CN(C)C([S-])=S.CN(C)C([S-])=S |
Chemical Identifiers
| 14484-64-1 | |
| 416.47 | |
| WHDGWKAJBYRJJL-UHFFFAOYSA-K | |
| 86289068 | |
| [Fe+3].CN(C)C([S-])=S.CN(C)C([S-])=S.CN(C)C([S-])=S |
| C9H18FeN3S6 | |
| MFCD00067269 | |
| fuklasin, trimanzone, cormate, fermacide, fermate, ferradow, hexaferb, hokmate, trifungol, ferbam | |
| iron(3+) tris((dimethylcarbamothioyl)sulfanide) |
Specifications
| 14484-64-1 | |
| C9H18FeN3S6 | |
| 25 g | |
| fuklasin, trimanzone, cormate, fermacide, fermate, ferradow, hexaferb, hokmate, trifungol, ferbam | |
| [Fe+3].CN(C)C([S-])=S.CN(C)C([S-])=S.CN(C)C([S-])=S | |
| 416.47 | |
| 416.47 | |
| Crystalline Powder |
| Black | |
| MFCD00067269 | |
| 2811 | |
| WHDGWKAJBYRJJL-UHFFFAOYSA-K | |
| iron(3+) tris((dimethylcarbamothioyl)sulfanide) | |
| 86289068 | |
| ≥97.0% (T) | |
| Iron(III) Dimethyldithiocarbamate |
Safety and Handling
EINECSNumber : (2)-1833
RTECSNumber : NO8750000
TSCA : Yes