missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Dimethyldithiocarbamate 97.0+%, TCI America™

Catalog No. D126725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D126725G 25 g
D1267500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D126725G Supplier TCI America Supplier No. D126725G

Chemical Identifiers

CAS 14484-64-1
Molecular Formula C9H18FeN3S6
Molecular Weight (g/mol) 416.47
MDL Number MFCD00067269
InChI Key WHDGWKAJBYRJJL-UHFFFAOYSA-K
Synonym fuklasin, trimanzone, cormate, fermacide, fermate, ferradow, hexaferb, hokmate, trifungol, ferbam
PubChem CID 86289068
IUPAC Name iron(3+) tris((dimethylcarbamothioyl)sulfanide)
SMILES [Fe+3].CN(C)C([S-])=S.CN(C)C([S-])=S.CN(C)C([S-])=S

Specifications

Color Black
Quantity 25 g
UN Number 2811
Formula Weight 416.47
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iron(III) Dimethyldithiocarbamate
EINECSNumber (2)-1833
RTECSNumber NO8750000
TSCA Yes