missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Chloride Hexahydrate, puriss. p.a., Honeywell Fluka™

Catalog No. p-7072005
Change view
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000837 1 kg
60045924 6 x 1 kg
6000839 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6000837 Supplier Honeywell Chemicals Supplier No. 312321KG
Only null left

Puriss. p.a., Reagent European Pharmacopoeia,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 1 kg
Linear Formula FeCl3 · 6H2O
UN Number 3260
Formula Weight 270.3g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Vapor Pressure 1mmHg (194°C)
Chemical Name or Material Iron(Iii) Chloride Hexahydrate, Reag. Ph. Eur.
Hazard Statement P280-P305 + P351 + P338
Precautionary Statement H290-H302-H315-H318
EINECSNumber 231-729-4
RTECSNumber NO5425000