missing translation for 'onlineSavingsMsg'
Learn More

Iron(Iii) Chloride Hexahydrate, ACS Reagent, Honeywell Fluka™

Catalog No. 6000831
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000831 500 g
6000830 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000831 Supplier Honeywell Chemicals Supplier No. 236489500G
Only null left

ACS Reagent, 97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 500 g
Linear Formula FeCl3 · 6H2O
UN Number 3260
Formula Weight 270.3g/mol
Percent Purity 97%
Grade ACS Reagent
Vapor Pressure 1mmHg (194°C)
Chemical Name or Material Iron(Iii) Chloride Hexahydrate
Hazard Statement P280-P305 + P351 + P338
Precautionary Statement H290-H302-H315-H318
EINECSNumber 231-729-4
RTECSNumber NO5425000