missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) chloride hexahydrate, 99+%, for analysis

Catalog No. p-43908
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC217091000 100 g Plastic bottle
AC217095000 500 g Plastic bottle
AC217090025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC217091000 Supplier Thermo Scientific Chemicals Supplier No. 217091000
Only null left

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid for analysis

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280.0°C to 285.0°C
Melting Point 37.0°C
Color Orange
Physical Form Adhering Crystalline Powder and Lumps
Quantity 100 g
Assay Percent Range 99.0 to 102% (Iodometry)
Linear Formula FeCl3·6H2O
Fieser 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158
Merck Index 15, 4046
Solubility Information Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether
Formula Weight 270.29
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Iron(III) chloride hexahydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa
RTECSNumber NO5425000

RUO – Research Use Only