Learn More
Iron(III) chloride, anhydrous, 98%
Description
1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Boiling Point | 316°C |
| Melting Point | 306°C |
| Color | Black |
| Physical Form | Crystalline |
| Quantity | 10 g |
| Assay Percent Range | 98% |
| Solubility Information | Freely soluble in methanol,ethanol,ether,and acetone. |
| Formula Weight | 162.21 |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H317-H318 |
| Precautionary Statement | P234-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 231-729-4 |
| RTECSNumber | LJ9100000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only