missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfide, 99.9% (metals basis)

CAS: 1317-37-9 | FeS | 87.91 g/mol

Supplier:  Thermo Scientific Chemicals 01402409

 View more versions of this product

Catalog No. AA1402409


Only null left
Explore available promotions

Description

Description

Iron(II) sulfide is used to generate hydrogen sulfide instantaneously by reacting with hydrochloric acid. In laboratory, Kipp’s apparatus is utilized for this purpose. It is used as a re-sulphurizing and alloying agent, as a reducing agent to remove heavy metal impurities from phosphoric acid. It is used to control hydrogen embrittlement in alloy and stainless steel industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron(II) sulfide
1317-37-9
Powder
99.9% (Metals basis)
MFCD00011013
Insoluble in water and nitric acid.
[S--].[Fe++]
87.91
87.91
Air and moisture sensitive
1195°C
Brown to Black
10 g
FeS
ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
GNVXPFBEZCSHQZ-UHFFFAOYSA-N
λ²-iron(2+) sulfanediide
14828
Odorless
4.74 g/cm3

Chemical Identifiers

1317-37-9
87.91
GNVXPFBEZCSHQZ-UHFFFAOYSA-N
14828
FeS
MFCD00011013
ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
[S--].[Fe++]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

This material and/or its container must be disposed of as hazardous waste.

Contact with acids liberates toxic gas.

EINECSNumber : 215-268-6

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only