missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfide, 99.9% (metals basis)

Catalog No. p-4781951
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1402403 1 g
AA1402409 10 g
AA1402418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1402403 Supplier Thermo Scientific Chemicals Supplier No. 01402403
Only null left

CAS: 1317-37-9 | FeS | 87.91 g/mol

Iron(II) sulfide is used to generate hydrogen sulfide instantaneously by reacting with hydrochloric acid. In laboratory, Kipp’s apparatus is utilized for this purpose. It is used as a re-sulphurizing and alloying agent, as a reducing agent to remove heavy metal impurities from phosphoric acid. It is used to control hydrogen embrittlement in alloy and stainless steel industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-37-9
Molecular Formula FeS
Molecular Weight (g/mol) 87.91
MDL Number MFCD00011013
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
PubChem CID 14828
SMILES [S--].[Fe++]

Specifications

Melting Point 1195°C
Color Brown to Black
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9% (Metals basis)
Solubility Information Insoluble in water and nitric acid.
IUPAC Name sulfanylideneiron
Formula Weight 87.91
Odor Odorless
Sensitivity Air and moisture sensitive
Density 4.74
Chemical Name or Material Iron(II) sulfide
Show More Show Less
Hazard Category Contact with acids liberates toxic gas.
Hazard Statement This material and/or its container must be disposed of as hazardous waste.
EINECSNumber 215-268-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only