missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7049121
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AA1296706 5 g
AA1296714 25 g
2 options

Catalog No. AA1296706

Supplier: Thermo Scientific Chemicals 01296706

Only null left

CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol

Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Melting Point 64°C (in -6H2O)
Color Green
Physical Form Crystalline
Quantity 5 g
Linear Formula FeSO4·7H2O
Merck Index 14,4057
Solubility Information Soluble in water. Insoluble in alcohol.
IUPAC Name iron(2+);sulfate;heptahydrate
Formula Weight 278.02 (151.91 Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Air Sensitive
Density 1.897 g/mL
Assay (metals basis)
Chemical Name or Material Iron(II) sulfate heptahydrate
Show More Show Less
Hazard Category H302-H315-H319-H336-H373
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
EINECSNumber 231-753-5
RTECSNumber NO8510000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only