missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) sulfate heptahydrate, 98%

CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol

Supplier:  Thermo Scientific Chemicals A151780C

 View more versions of this product

Catalog No. AAA151780C

Only null left
Explore available promotions

Description

Description

Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Air and moisture sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Iron(II) sulfate heptahydrate
7782-63-0
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron(2+);sulfate;heptahydrate
62662
278.02 (151.91 Anhydrous)
Air Sensitive
64°C (in -6H2O)
10,000 g
FeSO4·7H2O
14,4057
Soluble in water. Insoluble in alcohol.
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
278.01
CHEBI:75836
98%
1.897

Chemical Identifiers

7782-63-0
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
FeH14O11S
MFCD00149719
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H302-H315-H319-H336-H373

EINECSNumber : 231-753-5

RTECSNumber : NO8510000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only