missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) perchlorate hydrate, Reagent Grade

Catalog No. AA1164622
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1164622 100 g
AA1164636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1164622 Supplier Thermo Scientific Chemicals Supplier No. 01164622

CAS: 335159-18-7 | Cl2FeO8 | 254.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 335159-18-7
Molecular Formula Cl2FeO8
Molecular Weight (g/mol) 254.74
MDL Number MFCD00150234
InChI Key CKFMJXZQTNRXGX-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diperchlorate
SMILES [Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point >100°C (decomposition)
Physical Form Crystalline
Quantity 100 g
UN Number UN1481
Solubility Information Soluble in alcohol and water (?98g/100 mL H2O at 0?°C).
Formula Weight 254.75 (Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Iron(II) perchlorate hydrate
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-704-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only