accessibility menu, dialog, popup

UserName

OClOOOOClOOOFe

CAS RN 335159-18-7

IUPAC Name: λ²-iron(2+) diperchlorate
Synonyms: iron diperchlorate
Molecular Weight (g/mol): 254.74
Molecular Formula: Cl2FeO8
InChi Key: CKFMJXZQTNRXGX-UHFFFAOYSA-L
SMILES: [Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results