missing translation for 'onlineSavingsMsg'
Learn More

Iron(II,III) oxide, 97% (metals basis)

Catalog No. p-4407477
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12374A1 1 kg
AA12374A7 5 kg
AA12374C4 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA12374A1 Supplier Thermo Scientific Chemicals Supplier No. 012374A1
Only null left

CAS: 1317-61-9 | Fe3O4 | 231.53 g/mol

Mostly it is used as a black- pigment -catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning, whereas it is used as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymer, rubber, filler, building, construction, appliances, and magnetic inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1317-61-9
Molecular Formula Fe3O4
Molecular Weight (g/mol) 231.53
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
Synonym Ferrosoferric oxide, Ferric ferrous oxide
IUPAC Name diiron(3+) λ²-iron(2+) tetraoxidandiide
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]

Specifications

Melting Point 1538°C
Physical Form Powder
Quantity 1 kg
Assay Percent Range 97% (metals basis)
Solubility Information Insoluble in water and organic solvents. Soluble in concentrated mineral acids.
Formula Weight 231.54
Refractive Index 3
Density 5.2 g/mL
Chemical Name or Material Iron(II, III) oxide
EINECSNumber 215-277-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only