missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride hydrate, Reagent Grade, 99%

Catalog No. AA1248414
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1248414 25 g
AA1248430 250 g
AA12484A1 1 kg
AA12484A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1248414 Supplier Thermo Scientific Chemicals Supplier No. 01248414
Only null left

Air Sensitive | CAS: 23838-02-0 | Cl2Fe . x H2O | 126.75 g/mol

It is used as a reducing agent in metallurgy, in pharmaceutical preparations, as a mordant in dyeing and in sewage treatment. Ferrous chloride has a kind of niche application. During the laboratory synthesis of iron complexes, ferrous chloride acts as a reducing flocculating agent in wastewater treatment, particularly for wastes containing chromate. It is the precursor to hydrated iron(III) oxides that are magnetic pigments. Ferrous chloride is employed as a reducing agent in many organic synthesis reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23838-02-0
Molecular Formula Cl2Fe . x H2O
Molecular Weight (g/mol) 126.75
MDL Number MFCD00149711
InChI Key NMCUIPGRVMDVDB-UHFFFAOYSA-L
Synonym Iron dichloride, Iron protochloride
IUPAC Name λ²-iron(2+) dichloride
SMILES [Cl-].[Cl-].[Fe++]

Specifications

Color Green-Yellow
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99%
Solubility Information Soluble in water,alcohol and acetone. Insoluble in ether.
Formula Weight 126.75 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Air Sensitive
Chemical Name or Material Iron(II) chloride hydrate
Hazard Category H302-H314-H335
Hazard Statement Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-843-4
RTECSNumber NO5600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only