accessibility menu, dialog, popup

UserName

FeClCl

CAS RN 23838-02-0

IUPAC Name: λ²-iron(2+) dichloride
Synonyms: iron(II) chloride
Molecular Weight (g/mol): 126.75
Molecular Formula: Cl2Fe . x H2O
InChi Key: NMCUIPGRVMDVDB-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Fe++]
Grade 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)