Learn More
Iron(II) chloride, 97%, anhydrous
CAS: 7758-94-3 | Cl2Fe | 126.75 g/mol
$184.45 - $184.45
Chemical Identifiers
| CAS | 7758-94-3 |
|---|---|
| Molecular Formula | Cl2Fe |
| Molecular Weight (g/mol) | 126.75 |
| MDL Number | MFCD00011004 |
| InChI Key | NMCUIPGRVMDVDB-UHFFFAOYSA-L |
| Synonym | Ferrous chloride |
| IUPAC Name | λ²-iron(2+) dichloride |
| SMILES | [Cl-].[Cl-].[Fe++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389350250
|
Thermo Scientific Chemicals
389350250 |
25 g | Glass bottle |
Each for $184.45
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7758-94-3 | |
| 126.75 | |
| NMCUIPGRVMDVDB-UHFFFAOYSA-L | |
| λ²-iron(2+) dichloride |
| Cl2Fe | |
| MFCD00011004 | |
| Ferrous chloride | |
| [Cl-].[Cl-].[Fe++] |
Specifications
| 1026.0°C | |
| 7758-94-3 | |
| Powder | |
| 97% | |
| FeCl2 | |
| 15,407 | |
| Solubility in water: 644g/L | |
| [Cl-].[Cl-].[Fe++] | |
| 126.75 | |
| 97% | |
| Iron(II) chloride |
| 677.0°C | |
| Brown | |
| 25 g | |
| Cl2Fe | |
| MFCD00011004 | |
| Ferrous chloride | |
| NMCUIPGRVMDVDB-UHFFFAOYSA-L | |
| λ²-iron(2+) dichloride | |
| 126.75 | |
| Glass bottle |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
EINECSNumber : 231-843-4
RUO – Research Use Only