Learn More
Iron AA Standard, Certified, 1000ppm ±5ppm (1mL = 1mg Fe), LabChem™
Supplier: LabChem LC157001
Specifications
| Iron AA Standard | |
| Poly Bottle | |
| 96.42,2.86,0.72 | |
| 1g/mL | |
| Fe | |
| Fe(NO3)3·9H2O | |
| UN3264 | |
| Soluble in water | |
| [Fe] | |
| 55.85 | |
| 23925 | |
| 404.02 |
| 1000ppm (1mL = 1mg Fe) | |
| 7439-89-6,7782-61-8,7782-61-8,7697-37-2,7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±5 ppm (1mL = 1mg Fe) | |
| MFCD00010999 | |
| Colorless | |
| XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
| iron | |
| 500 mL | |
| CHEBI:82664 | |
| Certified |
Chemical Identifiers
| 7439-89-6 | |
| 55.85 | |
| XEEYBQQBJWHFJM-UHFFFAOYSA-N | |
| CHEBI:82664 | |
| [Fe] |
| Fe | |
| MFCD00010999 | |
| 23925 | |
| iron |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature