missing translation for 'onlineSavingsMsg'
Learn More

Iron 57, 57Fe, plasma standard solution, Specpure™, 57Fe 10μg/mL

 

Supplier:  Thermo Scientific Chemicals 045277AD

Catalog No. AA45277AD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Iron 57, 57Fe, plasma standard solution
Liquid
UN3264
Specpure
57Fe 10μg/mL
MFCD00144503
50 mL

Chemical Identifiers

MFCD00144503
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390

H290-H315-H319

DOTInformation : Hazard Class: 8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only