missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride trihydrate, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 461540010

Quantity 1g

 View more versions of this product

Catalog No. AC461540010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

136572-09-3
677.18
irinotecan hydrochloride trihydrate, irinotecan hcl trihydrate, cpt 11, unii-042laq1iis, campto, irinotecan hcl, 042laq1iis, onivyde, cpt-11
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.O.O.O.Cl
HCl·3H2O
KLEAIHJJLUAXIQ-JDRGBKBRSA-N
60837

Specifications

Irinotecan hydrochloride trihydrate
Authentic
1g
Solubility in water: .. Other solubilities: soluble in methanol
CCC1=C2C=C(C=CC2=NC3=C1CN4C3=CC5=C(C4=O)COC(=O)C5(CC)O)OC(=O)N6CCC(CC6)N7CCCCC7.O.O.O.Cl
60837
≥96.0% (HPLC)
136572-09-3
HCl·3H2O
irinotecan hydrochloride trihydrate, irinotecan hcl trihydrate, cpt 11, unii-042laq1iis, campto, irinotecan hcl, 042laq1iis, onivyde, cpt-11
KLEAIHJJLUAXIQ-JDRGBKBRSA-N
677.18
677.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions