accessibility menu, dialog, popup

UserName

CAS RN 136572-09-3

IUPAC Name: hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate trihydrate chloride
Synonyms: hydrogen irinotecan trihydrate chloride
Molecular Weight (g/mol): 677.19
Molecular Formula: C33H45ClN4O9
InChi Key: KLEAIHJJLUAXIQ-JDRGBKBRSA-N
SMILES: [H+].O.O.O.[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)