missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride trihydrate, 99%

Catalog No. p-7058221
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62370MC 100 mg
AAJ62370MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62370MC Supplier Thermo Scientific Chemicals Supplier No. J62370MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Topoisomerase I inhibitor | CAS: 136572-09-3 | C33H39ClN4O6 | 623.15 g/mol

Topoisomerase I inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136572-09-3
Molecular Formula C33H39ClN4O6
Molecular Weight (g/mol) 623.15
MDL Number MFCD01862255
InChI Key GURKHSYORGJETM-WAQYZQTGSA-N
Synonym irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe
PubChem CID 74990
ChEBI CHEBI:5971
IUPAC Name hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride
SMILES [H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O

Specifications

Melting Point 256°C
Color Yellow
Quantity 100 mg
Merck Index 14,5091
Solubility Information Soluble in DMSO at 100mg/ml; Soluble in water at 25mg/ml with warming
Formula Weight 677.19 (623.15 Anhydrous)
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Irinotecan hydrochloride trihydrate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber DW1060750
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only