Learn More
Irinotecan hydrochloride
Topoisomerase I inhibitor | CAS: 100286-90-6 | C33H39ClN4O6 | 623.15 g/mol
Supplier: Thermo Scientific Chemicals J60743MF
Description
Topoisomerase I inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Irinotecan hydrochloride | |
| Yellow | |
| MFCD01862255 | |
| 14,5091 | |
| Soluble in DMSO or DMF at approximately 20mg/ml; Sparingly soluble in aqueous buffers; /n | |
| [H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O | |
| 623.15 | |
| CHEBI:5971 | |
| Powder |
| 100286-90-6 | |
| C33H39ClN4O6 | |
| 50 mg | |
| irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe | |
| GURKHSYORGJETM-WAQYZQTGSA-N | |
| hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0â´,â¹.0¹âµ,²â°]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride | |
| 74990 | |
| 623.14 |
Chemical Identifiers
| 100286-90-6 | |
| 623.15 | |
| GURKHSYORGJETM-WAQYZQTGSA-N | |
| 74990 | |
| hydrogen (19S)-10,19-diethyl-19-hydroxy-14,18-dioxo-17-oxa-3,13-diazapentacyclo[11.8.0.0²,¹¹.0â´,â¹.0¹âµ,²â°]henicosa-1(21),2(11),3,5,7,9,15(20)-heptaen-7-yl [1,4'-bipiperidine]-1'-carboxylate chloride |
| C33H39ClN4O6 | |
| MFCD01862255 | |
| irinotecan hydrochloride, irinotecan hcl, topotecin, campto, cpt-11, cpt 11, camptothecin 11 hydrochloride, camptothecin 11, unii-06x131e4oe | |
| CHEBI:5971 | |
| [H+].[Cl-].CCC1=C2C=C(OC(=O)N3CCC(CC3)N3CCCCC3)C=CC2=NC2=C1CN1C2=CC2=C(COC(=O)[C@]2(O)CC)C1=O |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
RTECSNumber : DW1061000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only