missing translation for 'onlineSavingsMsg'
Learn More

Iridium standard solution, for AAS, 1 mg/ml Ir in 5% HCl, Thermo Scientific Chemicals

$141.33 - $141.33

Specifications

Chemical Name or Material Iridium standard solution
CAS 13569-57-8,7647-01-0,7732-18-5
Density 1.0200g/mL
Physical Form Solution
Molecular Formula Cl3H6IrO3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC196041000
View Documents
Thermo Scientific Chemicals
196041000
100 mL
Each for $141.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Iridium standard solution
1.0200g/mL
Cl3H6IrO3
Pink
O.O.O.Cl[Ir](Cl)Cl
352.612
AAS
13569-57-8,7647-01-0,7732-18-5
Solution
990 to 1010 μg/mL of Ir(3+) in 10 to 20% HCl
LNJXVUXPFZKMNF-UHFFFAOYSA-K
trichloroiridium;trihydrate
6102153
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only