missing translation for 'onlineSavingsMsg'
Learn More

Iridium(III) chloride, anhydrous, Premion™, 99.99% (metals basis), Ir 63.9% min

Catalog No. p-4408124
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4338006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4338006 Supplier Thermo Scientific Chemicals Supplier No. 04338006
Only null left

CAS: 10025-83-9 | Cl3Ir | 298.567 g/mol

Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10025-83-9
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.567
MDL Number MFCD00011063
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
PubChem CID 25563
IUPAC Name trichloroiridium
SMILES Cl[Ir](Cl)Cl

Specifications

Melting Point 763°C (decomposition)
Color Brown to Green
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 298.56
Sensitivity Hygroscopic
Appearance Green to brown powder
Chemical Name or Material Iridium(III) chloride
missing translation for 'healthHazard1' H290-H314-H335
missing translation for 'healthHazard3' P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
EINECSNumber 233-044-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only