missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium ICP Standard, 1000 ppm Ir in 10% HCl, Ricca Chemical
$817.82 - $817.82
Chemical Identifiers
| CAS | 13569-57-8 |
|---|---|
| Molecular Formula | Cl3H6IrO3 |
| Molecular Weight (g/mol) | 352.612 |
| InChI Key | LNJXVUXPFZKMNF-UHFFFAOYSA-K |
| PubChem CID | 6102153 |
| IUPAC Name | trichloroiridium;trihydrate |
| SMILES | O.O.O.Cl[Ir](Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PIR1KH-100
|
Ricca Chemical Company
PIR1KH100 |
100 mL | Natural LDPE Bottle |
Each for $817.82
|
|
||||
Chemical Identifiers
| 13569-57-8 | |
| 352.612 | |
| 6102153 | |
| O.O.O.Cl[Ir](Cl)Cl |
| Cl3H6IrO3 | |
| LNJXVUXPFZKMNF-UHFFFAOYSA-K | |
| trichloroiridium;trihydrate |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Ir | |
| 91.0,10.75,0.17 | |
| Liquid | |
| Brown | |
| Approximately 0°C | |
| LNJXVUXPFZKMNF-UHFFFAOYSA-K | |
| trichloroiridium;trihydrate | |
| 100 mL | |
| Iridium ICP Standard |
| 13569-57-8,7647-01-0,13569-57-8,7732-18-5 | |
| 88.0,10.33,0.16 | |
| 1 mL = 1 mg Ir | |
| Cl3H6IrO3 | |
| < 2 | |
| Standard | |
| O.O.O.Cl[Ir](Cl)Cl | |
| 352.612 | |
| 6102153 |