Learn More
Invitrogen™ IRF7 Monoclonal Antibody (MNGPKL), PE, eBioscience™, Invitrogen™
Description
Description: The monoclonal antibody MNGPKL reacts with mouse interferon regulatory factor 7 (IRF7), a 54 kDa transcription factor essential for induction of the antiviral innate immune response and IFN alpha production. B cells, plasmacytoid dendritic cells (pDC), and monocytes constitutively express IRF7 in the cytoplasm until activation by viral infection, double-stranded RNA or Toll-like receptor (TLR) signaling. CpG ODNs and single-stranded RNA, recognized by CD289 (TLR9) and TLR7, respectively, trigger IRF7 activation and upregulation. This activation promotes type I IFN response to disrupt pathogen and viral infection. These events cause IRF7 phosphorylation and translocation to the nucleus where it forms a homodimer to affect transcription. Applications Reported: This MNGPKL antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MNGPKL antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse splenocytes using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.
Specifications
Specifications
| Antigen | IRF7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MNGPKL |
| Concentration | 0.2 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | IRF7 |
| Gene Accession No. | P70434 |
| Gene Alias | IMD39; Interferon regulatory factor 7; interferon regulatory factor-7H; Irf7; IRF-7; IRF7A; IRF7B; IRF7C; IRF7H; IRF-7H |
| Show More |
For Research Use Only.