missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRF5 Monoclonal Antibody (K40049_14B6), Invitrogen™

Catalog No. MA555783
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555783 50 μL
MA556163 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA555783 Supplier Invitrogen™ Supplier No. MA555783
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: TGKYTEGVDE ADPAKWKANL RCALNKSRDF RLIYDGPRDM PPQPYKIYEV CSNGPAPTDS QPPEDYSFGA GEEEEEEEEL QRMLPSLSLT EDVKWPPTLQ

This gene encodes a member of the interferon regulatory factor (IRF) family, a group of transcription factors with diverse roles, including virus-mediated activation of interferon, and modulation of cell growth, differentiation, apoptosis, and immune system activity. Members of the IRF family are characterized by a conserved N-terminal DNA-binding domain containing tryptophan (W) repeats. Alternative splice variants encoding different isoforms exist.

Specifications

Antigen IRF5
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K40049_14B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IRF5
Gene Accession No. Q13568
Gene Alias AW491843; Interferon regulatory factor 5; interferon regulatory factor 5 bone marrow; IRF5; IRF-5; mirf5; OTTHUMP00000212907; SLEB10
Gene Symbols IRF5
Host Species Mouse
Immunogen Recombinant human IRF5 (immunogen range is 71-170).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3663
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less