missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRF4 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA5149021
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The IRF (interferon regulatory factor) family of transcription factors are characterized by an unique tryptophan pentad repeat DNA-binding domain. The IRFs are important in the regulation of interferons in response to infection by virus, and in the regulation of interferon-inducible genes. IRF4 is lymphocyte specific and negatively regulates toll-like-receptor (TLR) signaling that is central to the activation of innate and adaptive immune systems. A chromosomal translocation involving this gene and the IgH locus, t(6;14)(p25;q32), may be a cause of multiple myeloma. Alternatively spliced transcript variants have been found.Chemical Identifiers
| IRF4 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human | |
| IRF4 | |
| AI385587; interferon regulatory factor 4; Irf4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; multiple myeloma oncogene 1; MUM1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; SHEP8; Spip; Transcriptional activator PIP | |
| IRF4 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Sheep | |
| RUO | |
| PBS with trehalose and no preservative | |
| Q15306 | |
| 3662 | |
| E. coli-derived recombinant human IRF4, Glu130-Glu451. | |
| Antibody |
Specifications
| IRF4 | |
| Polyclonal | |
| Unconjugated | |
| IRF4 | |
| AI385587; interferon regulatory factor 4; Irf4; IRF-4; LSIRF; Lymphocyte-specific interferon regulatory factor; multiple myeloma oncogene 1; MUM1; NF-EM5; PU.1 interaction partner; Sfpi1/PU.1 interaction partner; SHEP8; Spip; Transcriptional activator PIP | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 3662 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q15306 | |
| IRF4 | |
| E. coli-derived recombinant human IRF4, Glu130-Glu451. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |